Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3-Dimethoxycyclobutane-1-carboxylate methyl ester

Supplier: MATRIX SCIENTIFIC MS

113056-1G 113056-5G
102897-322EA 40 USD InStock
102897-322 102897-324
3,3-Dimethoxycyclobutane-1-carboxylate methyl ester
3,3-Dimethoxycyclobutane-1-carboxylate methyl ester
Hazard: Irritant


Formula: C₈H₁₄O₄
MDL Number: MFCD11111944
CAS Number: 98231-07-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty