Ethyl-1-benzyl-4-oxo-3-piperidinecarboxylate ≥97%

Supplier: MATRIX SCIENTIFIC MS

114008-1G 114008-10G
102893-126EA 253.85 USD
102893-126 102893-128
Ethyl-1-benzyl-4-oxo-3-piperidinecarboxylate ≥97%
Ethyl-1-benzyl-4-oxo-3-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₉NO₃
MW: 261.32 g/mol
MDL Number: MFCD00792540
CAS Number: 41276-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR