PFA Tubing, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N8145239 N8145240 N8145242 N8145244
10139-628EA 287.54 USD
10139-628 10139-630 10139-632 10139-766 10139-768 10139-770 10139-772 10139-774
PFA Tubing, PerkinElmer
Tubing Transfer Tubing
Ultra-clean fittings and PFA tubing are ideal for all trace metal analysis applications.
  • Easy to use
  • Excellent clarity
  • Flexible
  • High continuous service temperature
Ferrules install quickly and easily for high-flow applications.