Tegaderm™ Hydrocolloid Dressing, 3M™
Supplier: Healthcare Products
MMM90007 MMM 90003 MMM 90004 MMM 90005 MMM 90001 MMM 90002
75787-454CS 700.85 USD InStock
75787-454 75787-754 75787-756 75787-772 75787-836 75787-840
Tegaderm™ Hydrocolloid Dressing, 3M™
First Aid Supplies Wound Dressings
Hydrocolloid dressing maintains an optimal moist wound environment which has been shown to enhance healing.
  • For low to moderate draining wounds
  • Significantly higher absorbency than leading hydrocolloid
  • Smooth outer surface helps reduce the risk of shear and friction
  • Breathable outer film layer provides consistently high rate of moisture vapor transmission
Available in oval, square, and sacral shapes. Film border on oval and sacral dressings eliminates the need for tape and makes dressing application easier.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty