(7-Hydroxy[1,2,4]triazolo[1,5-a]pyrimidin-6-yl)acetic acid

Supplier: MATRIX SCIENTIFIC MS

077533-500MG 077533-1G 077533-5G
102666-746EA 516.92 USD
102666-746 102666-748 102666-750
(7-Hydroxy[1,2,4]triazolo[1,5-a]pyrimidin-6-yl)acetic acid
(7-Hydroxy[1,2,4]triazolo[1,5-a]pyrimidin-6-yl)acetic acid

Hazard: Irritant

Formula: C₇H₆N₄O₃
MW: 194.15 g/mol
MDL Number: MFCD20441367
CAS Number: 1338495-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR