EC Broth, Tubes, Northeast Lab Services
Supplier: HERAEUS SILICA & METALS LTD
T1455 T1456
101374-568PK 138.4 USD InStock
101374-568 101374-570
EC Broth, Tubes, Northeast Lab Services
Media for Microbiology Dehydrated Media for Microbiology
EC Broth contains a convenient Durham Tube.
  • Glass Screw Tops
  • Prepared Medium
  • Durham Tubes Included
  • Fill Volume: 9.9 mL or 10 mL
  • Size: 16 mm X 125 mm
Two fill volumes are available to meet different lab needs.
NEL produces media tubes for a wide variety of clients including private laboratories, food processors, clinics and hospitals, academic institutions, and government agencies.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty