Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

XLT 4 Agar, Plates, Northeast Lab Services

Supplier: HERAEUS SILICA & METALS LTD

P1751 P1750
101374-182PK 39.08 USD InStock
101374-182 101374-180
XLT 4 Agar, Plates, Northeast Lab Services
Media for Microbiology Dehydrated Media for Microbiology
XLT 4 Agar is available with nalidixic acid.

  • Standard Petri Dish
  • Nalidixic Acid Available
  • Prepared Medium
  • Fill Volume: 20 mL
  • Size: 15 mm x 100 mm

All formulations are contained in a 100 mm x 15 mm standard petri dish.

They have a fill volume of 20 mL

Certificaciones: ISO Class 5 / FS209E Class 100 Validated EC Rooms certified.


Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty