Vee Gee Volumetric Flasks with Glass Stopper
Supplier: Heathrow Scientific
2306A-1000PK 2306A-50PK
490011-058PK 105.28 USD
490011-058 490011-062
Vee Gee Volumetric Flasks with Glass Stopper
Flasks Volumetric Flasks
These volumetric flasks feature machine-blown bodies and heavy-beaded, heavy-tubing necks that are tooled for standard taper glass stoppers. Our flasks are constructed of premium borosilicate glass, ensuring exceptional clarity and resistance to chemical interactions.
  • Made of premium borosilicate glass with a low coefficient of expansion
  • Designed 'to contain' with a temperature of calibration at 68 °F (20 °C)
  • The graduation line is sharply defined and of uniform width
  • Each flask has a large marking square and includes a glass stopper
  • Wide range of sizes available
Each volumetric flask is supplied with a standard taper glass stopper which provides a leakproof seal. This airtight seal not only preserves the integrity of your samples but also enhances the reliability of your volumetric measurements.
Certificaciones: Meets ASTM E-288 Class A specifications & USP standards for glassware