Agar Powder, Bacteriological Grade, HiMedia
Supplier: PAESEL+LOREI GMBH&CO
GRM026-500G GRM026-5KG GRM026-2.5KG GRM026-25KG
95022-838EA 254.32 USD
95022-838 95043-010 89132-792 97068-678 89400-370
Agar Powder, Bacteriological Grade, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Agar powder is specifically produced for use in bacteriological culture media and plant tissue culture media.