SHOWA 282 TEM-RES Insulated Cold Protective Glove, Showa

Supplier: Showa
282L-09 282M-08 282XL-10
89217-722CS 3968.43 USD
89217-722 89217-720 89217-724
SHOWA 282 TEM-RES Insulated Cold Protective Glove, Showa
Gloves
Keep hands warm and dry in temperatures as low as -31°C (-25°F)

  • Coating creates impenetrable barrier, keeping hands dry and warm
  • Breathable coating reduces perspiration
  • Polyurethane coating provides excellent wet/dry grip

Waterproof yet breathable, SHOWA 282 gloves use thermal insulation and state of the art TEM-RES technology to keep hands warm, dry, and well-protected in cold climates.

Gloves maintain flexibility in extremely low temperatures.

Información para pedidos: Order by Master case of 12 pair.

Envase: Packaged 12 pair per Master case. Each pair is individually poly bagged.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR