Ruthenium 1% on carbon

Supplier: Origen Biomedical, Inc

047296-14 047296-22
AA47296-14EA 240 USD
AA47296-14 AA47296-22
Ruthenium 1% on carbon
Ruthenium

4 to 8 mesh granular

Formula: Ru
MW: 101.07
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR