All-trans-retinal

Supplier: SPECTRUM CHEMICAL MFG CORP

R3041-100MG R3041-250MG R3041-5GM R3041-1GM
700003-794EA 195.4 USD
700003-794 700003-798 700003-800 700003-796
All-trans-retinal
All-trans-retinal

all-trans-Retinal is also known as retinaldehyde and considered a form of vitamin A. It is the chemical basis of animal vision.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₀H₂₈O
MW: 284.4357 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00001550
CAS Number: 116-31-4

Order Now

Specification Test Results

Appearance Yellow Powder
Melting Point 57 - 65°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR