Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Ethyldiisopropylamine ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

D2163-100ML D2163-4LT D2163-500MLGL
10841-902EA 98.77 USD InStock
10841-902 10841-904 10841-906
N-Ethyldiisopropylamine ≥99%
N-Ethyldiisopropylamine
N,N-Diisopropylethylamine is a colorless liquid and is also known as Hunig's base. N,N-DIISOPROPYLETHYLAMINE is a good base but a poor nucleophile. This characteristics make this material an extremely versatile synthetic organic reagent

GHS hazard : Flame;Corrosion;Skull and crossbones;Exclamation mark
UN2733


Formula: C₈H₁₉N
Density: 0.742
Storage Temperature: Ambient
MDL Number: MFCD00008868
CAS Number: 7087-68-5
UN: 2733
ADR: 38,II
Order Now
Specification Test Results
Assay 99 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty