Orange oil, sweet

Supplier: SPECTRUM CHEMICAL MFG CORP

OR115-125ML OR115-500MLG
700010-374EA 85.1 USD
700010-374 700010-378
Orange oil, sweet
Orange oil

Orange Oil, Sweet is created by cells within the rind of the orange and is an essential oil. It is often used to replace pure d-limonene which can be extracted from the oil through distillation. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.


GHS hazard : Flame;Exclamation mark
UN1197

Storage Temperature: Ambient
MDL Number: MFCD00147798
CAS Number: 8008-57-9
UN: 1197
ADR: 3 III

Order Now

Specification Test Results

Specific Gravity 0.840 - 0.846


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR