Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tozasertib ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

SYN-1092-M001 SYN-1092-M005 SYN-1092-M010 SYN-1092-M050
102991-980EA 74.77 USD InStock
102991-980 102991-982 102991-984 102991-986
Tozasertib ≥95%
Tozasertib
Tozasertib, or VX-680, is a potent and selective inhibitor of Aurora kinases, particularly Aurora A and B. In vitro, VX-680 blocks cell cycle progression and induces apoptosis in a wide range of human tumor types at low IC(50) values (i.e. 3.38nM for human BE-13 cells, and 14.34nM for NTERA cells). Moreover, VX680, also has very potent Ki values, with inhibition constants (Ki) of 0.6, 18, and 46nM for Aurora A, B, and C, respectively. VX-680 is also effective in vivo, being used in a Caki-1 xenograft model, VX-680 demonstrated a 75.7% (P < 0.001) decrease in Caki-1 xeno-graft tumor volume with no apparent alternation in animal body weight, peripheral blood counts, or other biological parameters.

In mammals, three Aurora kinases, Aurora A, Aurora B, and Aurora C, are expressed. Aurora C expression is mainly restricted to testis, where it functions during meiosis, whereas Aurora A and B play key functions regulating mitosis. Aurora A is localized to the centrosome and spindle poles, where it drives centrosome maturation, separation, and bipolar spindle assembly. Aurora B is the central component of the chromosomal passenger complex (CPC) that also contains the inner centromere protein (INCENP), borealin, and survivin. These non-enzymatic components of the CPC play a key role in regulating kinase activity and localization. Over-expression of Aurora kinases can be oncogeneic and they have thus long been targets for cancer thearpy.


Formula: C₂₃H₂₈N₈OS
Storage Temperature: Freezer
MDL Number: MFCD10565966
CAS Number: 639089-54-6
Order Now
Specification Test Results
Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty