Boc-Leu-Arg-Arg-AMC
Supplier: Van Der Voort Lucht en Milieu Amste
AG-CP3-0014-M001 AG-CP3-0014-M005
102988-474EA 41.54 USD InStock
102988-474 102988-476
Boc-Leu-Arg-Arg-AMC
Proteins and Peptides
Fluorogenic tri-peptide substrate for measuring the trypsin-like peptidase activity of the 20S proteasome. Substrate for the Kex2 endopeptidase from S. Cervisiae, which has substrate specificity toward the C-terminal side of LR, PR and RR sequences. Can be cleaved by Kallikrein 5 with a P1 site preference for basic residues (R and K). Excitation: 380nm. Emission: 460nm. This substrate is useful for inhibitor screening and kinetic analysis.
  • Soluble in 5% acetic acid.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty