Anthracene-9,10-dipropionic acid disodium salt

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

CDX-A0116-M010 CDX-A0116-M100
102989-188EA 274.15 USD
102989-188 102989-190
Anthracene-9,10-dipropionic acid disodium salt
Anthracene-9,10-dipropionic acid disodium salt

Water-​soluble naphthalene as singlet oxygen sensitizers. Self-​assembly building block.

Formula: C₂₀H₁₆Na₂O₄
MW: 366.32 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00467617
CAS Number: 82767-90-6

Order Now

Specification Test Results

Source/Host Synthetic.
Appearance/Color Yellow needles.
Solubility Soluble in methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR