Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenylpropionyl chloride ≥97%

Supplier: Van Der Voort Lucht en Milieu Amste
Danger

CDX-P0136-G001 CDX-P0136-G005
102990-876EA 84.46 USD InStock
102990-876 102990-878
2-Phenylpropionyl chloride ≥97%
2-Phenylpropionyl chloride
Building block for synthesis.


Formula: C₉H₉ClO
MW: 168.6 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03424723
CAS Number: 22414-26-2
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Source/Host Synthetic.
Purity 97% (GC)
Appearance/Color Colorless to light yellow liquid.
Solubility Soluble in chloroform.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty