Motesanib ≥95%

Supplier: Van Der Voort Lucht en Milieu Amste
Warning

SYN-1055-M001 SYN-1055-M005 SYN-1055-M010 SYN-1055-M050
102991-676EA 96.92 USD
102991-676 102991-678 102991-680 102991-682
Motesanib ≥95%
Motesanib
Formula: C₂₂H₂₃N₅O
MW: 373.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD10567689
CAS Number: 453562-69-1

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR