Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Makisterone A ≥95% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste

AG-CN2-0073-M001 AG-CN2-0073-M005 AG-CN2-0073-C250
102987-238EA 138.46 USD InStock
102987-238 102987-240 102987-964
Makisterone A ≥95% (by HPLC)
Makisterone A
A member of the ecdysteroid family. Ecdysone receptor (EcR) agonist. Induces the expression of genes coding for proteins that the larva requires, and it causes chromosome puffs (sites of high expression) to form in polytene chromosomes. Plays a role in insect development, cell proliferaton, growth and apoptosis by controlling gene expression involved in moulting and metamorphosis. It acts through a heterodimeric receptor comprising the ecdysone receptor and the ultraspiracle proteins (USP). Appears in plants mostly as a protection agent (toxins or antifeedants) against herbivorous insects. Could be used for controlled gene expression in scientific research, agriculture and medicine. Could be used for the development of selective insect growth regulators for use as environmentally benign insecticides.


Formula: C₂₈H₄₆O₇
Storage Temperature: Freezer
CAS Number: 20137-14-8
Order Now
Specification Test Results
Source/Host Isolated from Ipomoea hederacea.
Purity 95% (HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in acetic acid, ethanol, methanol or DMSO. Sparingly soluble in chloroform. Insoluble in water.

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty