Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

NS 1738 ≥99% (by HPLC)

Supplier: SAFE AND SURE NEDERLAND B.V.

CHM117-0010 CHM117-0050
76002-980EA 177.67 USD InStock
76002-980 76002-982
NS 1738 ≥99% (by HPLC)
NS 1738
A selective positive allosteric modulator selective for α7 nicotinic acetylcholine receptors (α7 nAChR).

NS 1738 exhibits cognitive enhancing effects in vivo.

Soluble in DMSO or ethanol.

Synonyms: 1-(5-chloro-2-hydroxyphenyl)-3-(2-chloro-5-(trifluoromethyl)phenyl)urea, α7 nAChR positive allosteric modulator


Formula: C₁₄H₉Cl₂F₃N₂O₂
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
MDL Number: MFCD11100189
CAS Number: 501684-93-1
Order Now
Specification Test Results
Purity ≥99% (HPLC)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty