Armenian Hamster IgG Isotype Control

Supplier: Bio X Cell
BE0091-25MG BE0091-50MG BE0091-100MG BE0091-FAB-5MG BE0091-1MG BE0091-5MG
103252-590EA 3920 USD
103252-590 103252-592 103252-594 103252-596 103252-600 103572-790 76555-090 76555-248 76555-292 76555-372 76555-444 76555-962
Armenian Hamster IgG Isotype Control
Antibodies
The polyclonal Armenian hamster IgG is purified from Armenian hamster serum. It is ideal for use as a non-reactive control IgG for Armenian hamster antibodies in most in vivo and in vitro applications.

Type: Control
Antigen:
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR