Yeast Morphology Agar, HiMedia Laboratories
Supplier: PAESEL+LOREI GMBH&CO
M138-100G
M138-500G
61000-020EA
138.98
USD
61000-020
95021-054
Yeast Morphology Agar, HiMedia Laboratories
Media for Microbiology
Dehydrated Media for Microbiology
Yeast Morphology Agar is recommended for use in the classification of yeasts on the basis of their colonial characteristics and cell morphology.
- Final pH (at 25 °C) is 5.6 ± 0.2
- Cream to yellow homogeneous free flowing powder