Anti-PD-1 (CD279) Armenian Hamster Monoclonal Antibody [clone: J43]

Supplier: Bio X Cell
BE0033-2-5MG BE0033-2-50MG BE0033-2-100MG BE0033-2-25MG BE0033-2-1MG
103252-438EA 3920 USD
103252-438 103252-440 103252-442 103252-444 103572-466 76555-152 76555-766 76555-976 76556-324 76557-234
Anti-PD-1 (CD279) Armenian Hamster Monoclonal Antibody [clone: J43]
Antibodies
InVivoMab anti-mouse PD-1 (CD279), Clone: J43, Isotype: Armenian hamster IgG, Applications: in vivo blocking of PD-1/PD-L signaling, in vitro PD-1 neutralization

Type: Primary
Antigen: PD-1 (CD279)
Clonality: Monoclonal
Clone: J43
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR