Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-MHC Class II (I-Ek/RT1-D) Mouse Monoclonal Antibody [clone: 14-4-4S (HB-32)]

Supplier: Bio X Cell
BE0167-1MG BE0167-100MG BE0167-50MG BE0167-25MG BE0167-5MG
103253-864EA 3920 USD InStock
103253-864 103253-866 103253-868 103253-870 103572-856 76554-524 76554-718 76555-908 76555-990 76556-004
Anti-MHC Class II (I-Ek/RT1-D) Mouse Monoclonal Antibody [clone: 14-4-4S (HB-32)]
Antibodies
InVivoMab anti-mouse/rat MHC Class II (I-Ek/RT1-D), Clone: 14-4-4S (HB-32), Isotype: mouse IgG2a, κ, Applications: in vivo blocking of antigen presentation, Flow cytometry

Type: Primary
Antigen: MHC Class II (I-Ek/RT1-D)
Clonality: Monoclonal
Clone: 14-4-4S (HB-32)
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty