Anti-4-1BB (CD137) Syrian Hamster Monoclonal Antibody [clone: 17B5]

Supplier: Bio X Cell
BE0296-100MG BE0296-50MG BE0296-25MG BE0296-1MG BE0296-5MG
103479-678EA 3920 USD
103479-678 103479-680 103479-682 103479-684 103572-658 76554-306 76554-736 76555-034 76555-294 76556-764
Anti-4-1BB (CD137) Syrian Hamster Monoclonal Antibody [clone: 17B5]
Antibodies
InVivoMAb anti-mouse 4-1BB (CD137), Clone: 17B5, Isotype: Syrian hamster IgG, Applications: in vitro 4-1BB blockade, Flow cytometry

Type: Primary
Antigen: 4-1BB (CD137)
Clonality: Monoclonal
Clone: 17B5
Conjugation:
Epitope:
Host: Syrian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR