Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acetic acid, sodium salt, [2-14C] ≥98%, MORpure™

Supplier: BMT - Dr.Brandt Medizin Technik

MC213-50UCI MC213-250UCI MC213-1MCI MC213-5MCI
101971-044EA 653.85 USD InStock
101971-044 101971-046 101971-048 101971-050
Acetic acid, sodium salt, [2-14C] ≥98%, MORpure™
Acetic acid, sodium salt, [2-14C]


CAS Number: 993-05-5
UN: 2910
ADR: ,
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty