Phenol, [14C(U)] ≥97%

Supplier: BMT - Dr.Brandt Medizin Technik

MC235-50UCI MC235-250UCI MC235-1MCI
101971-466EA 5387.69 USD
101971-466 101971-468 101971-470
Phenol, [14C(U)] ≥97%
Phenol, [14C(U)]
CAS Number: 53379-77-4
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR