Magnesium sulfate 3 M

Supplier: QGT Quarzglastechnik GmbH & Co KG

1008362 1008363
101443-380EA 102 USD
101443-380 101443-738
Magnesium sulfate 3 M
Magnesium sulphate solutions

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: MgSO₄·7H₂O
MW: 246.48 g/mol
MDL Number: MFCD00149785
CAS Number: 10034-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR