Static Dissipative, Flame Resistant/ARC Rated High Visibility Vest, Lakeland Industries
Supplier: Lakeland
V10AM0322ZL-MD V10AM0322ZL-XL V10AM0322ZL-3X
75923-614EA 161.76 USD
75923-614 75923-616 75923-618
Static Dissipative, Flame Resistant/ARC Rated High Visibility Vest, Lakeland Industries
Workwear
High Visibility Vest of Flame Resistant/ARC rated modacrylic mesh fabric to keep wearers visible in potentially hazardous atmospheres.
  • Static dissipative mesh fabric
  • Non-conductive zipper closure
Designed for industries where there is always concern for static build up, this material contains a carbon core fiber that will dissipate potentially harmful static charges. The Modacrylic Mesh fabric will never lose its FR and ARC rated protective properties. The vest features a non-conductive zipper front closure, 3 M Scotchlite reflective material, and carries an ATPV value of 5.1 Cal/cm2.
Certificaciones: ANSI 107-Class 2. ASTM F1506.