Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Coenzyme A trilithium salt dihydrate ≥75% (by enzymatic purity)

Supplier: THERMO SCIENTIFIC CHEMICALS INC MS

J13785-ME J13785-MC J13785-03 J13785-MD
101175-462EA 794.48 USD InStock
101175-462 101169-788 101170-786 101172-334
Coenzyme A trilithium salt dihydrate ≥75% (by enzymatic purity)
Coenzyme A trilithium salt dihydrate


Formula: C₂₁H₃₃N₇O₁₆P₃SLi₃·2H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00075848
CAS Number: 18439-24-2
EINECS: 242-317-9
Order Now
Specification Test Results
Assay ≥75% (Enzymatic)
Water Content (Karl Fischer Titration) ≤8%
Spectral Ratio (pH 7.5): A₂₅₀/A₂₆₀ 0.78 ±0.03
Spectral Ratio (pH 7.5): A₂₈₀/A₂₆₀ 0.16 ±0.03
Trace Metal, Lithium (Li) 2.5 ±1.5%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty