Anti-MUC1 Mouse Monoclonal Antibody (CF568) [clone: GP1.4]

Supplier: Biotium
BNC680159-100 BNC680159-500
75963-486EA 205.77 USD
75963-486 75963-488
Anti-MUC1 Mouse Monoclonal Antibody (CF568) [clone: GP1.4]
Antibodies
In Western blotting, this antibody recognizes proteins in MW range of 265-400 kDa, identified as different glycoforms of EMA. This MAb reacts with the DTRP epitope in the tandem repeats. Thealpha subunit has cell adhesive properties. It can act both as an adhesion and an anti-adhesion protein. EMA may provide a protective layer on epithelial cells against bacterial and enzyme attack. The beta subunit contains a C-terminal domain, which is involved in cell signaling, through phosphorylations and protein-protein interactions. In immunohistochemical assays, it superbly stains routine formalin/More Product Information
Order Now

SPECIFICATIONS

Antigen Symbol MUC1
Environmentally Preferable
Antigen Name Mucin 1 / EMA / Episialin / CD227
Antibody Type Primary
Clonality Monoclonal
Conjugation CF® 568
Clone GP1.4
Reactivity Human
Host Mouse
Gene ID Unigene ID: 89603
Isotype IgG1 kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage Buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR