Anti-THY1 Mouse Monoclonal Antibody [clone: F15-42-1]

Supplier: Biotium
BNUB0194-100 BNUB0194-500
75908-580EA 240.92 USD
75908-580 75908-638
Anti-THY1 Mouse Monoclonal Antibody [clone: F15-42-1]
Antibodies
CD90(Thy-1) is an 18-35 kDa GPI-anchored plasma membrane glycoprotein expressed in many cell types, such as in hematopoietic cells and neurons, connective tissues, various fibroblast and stromal cell lines, tumor endothelial cell lines and others. It is involved in T-cell activation, cellular adhesion, proliferation and migration, neurite outgrowth, wound healing, apoptosis, and fibrosis. CD90 participates in multiple signaling cascades and its effects are tissue- and cell type-specific. It often functions as an important regulator of cell-cell and cell-matrix interactions.

Type: Primary
Antigen: THY1
Clonality: Monoclonal
Clone: Clone F15-42-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR