Anti-CA9 Mouse Monoclonal Antibody (CF594) [clone: PN-15]

Supplier: Biotium
BNC940637-100 BNC940637-500
75971-208EA 205.77 USD
75971-208 75971-210
Anti-CA9 Mouse Monoclonal Antibody (CF594) [clone: PN-15]
Antibodies
This antibody recognizes a glycoprotein of ~200 kDa, identified as carbonic anhydrase IX (CAIX/gp200). Its epitope resides in the carbohydrate domain of gp200. It shows no significant cross-reactivity with other carbohydrate determinants, such as the Lewis blood group antigens, epithelial membrane antigen, HMFG, and AB blood group antigens. In normal kidney, gp200 is localized along the brush border of the pars convoluta and pars recta segments of the proximal tubule, as well as focally along the luminal surface of Bowman's capsule adjoining the outgoing proximal tubule. Reportedly, gp200 is expressed by 93% of primary and 84% of metastatic renal cell carcinomas. This MAb may be useful in the investigations of carcinomas of proximal nephrogenic differentiation especially those showing tubular differentiation.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®594 is a deep red fluorescent dye (Ex/Em 593/614 nm). It yields the brightest conjugates among spectrally similar dyes, and has excellent photostability.

Type: Primary
Antigen: CA9
Clonality: Monoclonal
Clone: Clone PN-15
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Horse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR