Anti-IGHM Mouse Monoclonal Antibody (CF594) [clone: DA4-4]
Supplier: Biotium
BNC940759-100 BNC940759-500
75953-280EA 205.77 USD
75953-280 75953-282
Anti-IGHM Mouse Monoclonal Antibody (CF594) [clone: DA4-4]
Antibodies
This antibody recognizes a protein of 75 kDa, identified as mu heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), gamma (IgG), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This MAb is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
CF® dyes areMore Product Information