Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-UCHL1 Mouse Monoclonal Antibody (CF594) [clone: UCHL1/775]

Supplier: Biotium
BNC940775-100 BNC940775-500
75969-876EA 205.77 USD InStock
75969-876 75969-878
Anti-UCHL1 Mouse Monoclonal Antibody (CF594) [clone: UCHL1/775]
Antibodies
This MAb reacts with a protein of 20-30 kDa, identified as PGP 9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UCHL-1). Initially, PGP 9.5 expression in normal tissues was reported in neurons and neuroendocrine cells but later it was found in distal renal tubular epithelium, spermatogonia, Leydig cells, oocytes, melanocytes, prostatic secretory epithelium, ejaculatory duct cells, epididymis, mammary epithelial cells, Merkel cells, and dermal fibroblasts. Furthermore, immunostaining for PGP 9.5 has been shown in a wide variety of mesenchymal neoplasms as well. A mutation in PGP 9.5 gene is believed to cause a form of Parkinson's disease.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®594 is a deep red fluorescent dye (Ex/Em 593/614 nm). It yields the brightest conjugates among spectrally similar dyes, and has excellent photostability.

Type: Primary
Antigen: UCHL1
Clonality: Monoclonal
Clone: Clone UCHL1/775
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty