Anti-IFNA1 Mouse Monoclonal Antibody (CF568) [clone: 2.52]

Supplier: Biotium
BNC680998-100 BNC680998-500
75952-016EA 205.77 USD
75952-016 75952-018
Anti-IFNA1 Mouse Monoclonal Antibody (CF568) [clone: 2.52]
Antibodies
This MAb is specific for human interferon alpha 1 and does not cross react with human interferon alpha 2. Interferons are widely used therapeutic agents because of their anti-tumor and anti-viral effects and because of their modulatory effects on the immune system. These cytokines produce their effects by binding to the Type 1 Interferon- & Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is influenced by phosphorylation of Serine 535 and 539 in the IFNAR1.

CF® dyes are Biotium's next-More Product Information
Order Now

SPECIFICATIONS

Antigen Symbol IFNA1
Environmentally Preferable
Antigen Name Interferon alpha 1
Antibody Type Primary
Clonality Monoclonal
Conjugation CF® 568
Clone 2.52
Reactivity Human
Host Mouse
Gene ID Unigene ID: 37026 , 533471
Isotype IgG1 kappa
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage Buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR