MAO-Glo Assay, Promega
Supplier: EBRO Electronic GmbH
V1401 V1402
PAV1401EA 782.49 USD
PAV1401 PAV1402
MAO-Glo Assay, Promega
Assays Cellular Assays
The MAO-Glo Assay provides a homogeneous luminescent method for measuring monoamine oxidase activity from recombinant and native sources and for testing the effects of test compounds on MAO activity.
  • Sensitive Detection of Monoamine Oxidase (MAO) Activity
  • Luminescence-based method eliminates need for time-consuming analyses such as HPLC
  • Simple "add and read" protocol ideal for high-throughput screening
  • Less MAO enzyme required than typical HPLC or fluorometric methods
The MAO-Glo Assay provides a homogeneous luminescent method for measuring monoamine oxidase (MAO) activity from recombinant and native sources and for testing the effects of test compounds on MAO activity. The MAO-Glo Assay is performed by incubating the MAO enzyme source with a luminogenic MAO substrate. The substrate of the MAO-Glo Assay is a derivative of beetle luciferin. Upon reaction with MAO, the derivative is converted into luciferin, which in turn reacts with luciferase to produce light. The amount of light produced is directly proportional to the activity of MAO. After the MAO reaction has been performed, the reconstituted Luciferin Detection Reagent is added. The reagent simultaneously stops the MAO reaction and initiates a stable glow-type luminescent signal with a half-life greater than 5 hours. This eliminates the need for strictly timed luminescent detection. The MAO-Glo Assay includes a luminogenic MAO substrate, two MAO Reaction Buffers (one that can be used with either MAO A or MAO B enzyme and one that is designed specifically for MAO B), a lyophilized Luciferin Detection Reagent and the Luciferin Detection Buffer. The user supplies the sample material containing MAO. Protocols are configured for multiwell plate formats but easily can be adapted for single-tube applications.