Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N4-Acetylsulfadimethoxine (synthetic) ≥98% (by HPLC)

Supplier: Van Der Voort Lucht en Milieu Amste

CDX-A0298-M025 CDX-A0298-M100
102989-288EA 279.69 USD InStock
102989-288 102989-290
N4-Acetylsulfadimethoxine (synthetic) ≥98% (by HPLC)
N4-Acetylsulfadimethoxine
Off-white powder

Soluble in chloroform


Storage Temperature: Freezer
CAS Number: 555-25-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty