Polysiloxane thio-functionalised, macroporous (metal absorbing resin) (Deloxan® THP II)
14-1870-50G 14-1870-10G 14-1870-250G
100197-378EA 653.91 USD InStock
100197-378 101324-542 100197-376
Polysiloxane thio-functionalised, macroporous (metal absorbing resin) (Deloxan® THP II)
Polysiloxane thio-functionalised
Deloxan® THP II Macroporous, Thiourea-Functionalized Polysiloxane (Metal Absorbing Resin)
Color and Form: Abrasion-proof spheres; Water content: 50-70%
Physical Characteristics: surface area 300-450m2/g (BET), pore volume 2.5-3.5 ml/g, density 0.55-0.65 kg/L (wet form)
Sold in collaboration with Evonik for research purposes only.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty