Chloro(di-2-norbornylphosphino)(2-dimethylaminomethylferrocen-1-yl)palladium(II) ≥97%

Warning

46-0272-250MG 46-0272-1G
100203-190EA 167.02 USD
100203-190 100203-192
Chloro(di-2-norbornylphosphino)(2-dimethylaminomethylferrocen-1-yl)palladium(II) ≥97%
Chloro(di-2-norbornylphosphino)(2-dimethylaminomethylferrocen-1-yl)palladium(II)
MDL Number: MFCD05861622
CAS Number: 614753-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR