Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Pentamethylcyclopentadienyl)tantalum tetrachloride ≥98%

Danger

73-0900-5G 73-0900-1G
100198-030EA 642.58 USD InStock
100198-030 100202-510
(Pentamethylcyclopentadienyl)tantalum tetrachloride ≥98%
(Pentamethylcyclopentadienyl)tantalum tetrachloride
Metallocenes, Derivatives & Cp Precursors


Formula: C₁₀H₁₅Cl₄Ta
Melting Pt: 220 °C
MDL Number: MFCD00070463
CAS Number: 71414-47-6
UN: 1759
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty