Palladium(II) sulphate

Danger

93-4609-1G 93-4609-5G
100201-112EA 174.09 USD
100201-112 100204-332
Palladium(II) sulphate
Palladium(II) sulphate
Formula: O₄PdS
MW: 202.48
MDL Number: MFCD00011173
CAS Number: 13566-03-5
UN: 3260
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR