Neodymium(III) isopropoxide ≥99.9% (Nd basis)

Danger

93-6012-1G 93-6012-5G
100207-882EA 83.51 USD
100207-882 100198-424
Neodymium(III) isopropoxide ≥99.9% (Nd basis)
Neodymium(III) isopropoxide

Alkoxide

Formula: C₉H₂₁NdO₃
MW: 321.503 g/mol
MDL Number: MFCD00058819
CAS Number: 19236-15-8
UN: 3178
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR