Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dihydrogen hexachloroosmate(IV) hexahydrate

Warning

93-7610-1G 93-7610-5G
100200-206EA 360.92 USD InStock
100200-206 100197-000
Dihydrogen hexachloroosmate(IV) hexahydrate
Dihydrogen hexachloroosmate(IV) hexahydrate


Formula: H₂Cl₆Os·6H₂O
MDL Number: MFCD00149908
CAS Number: 27057-71-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty