Dimethylzinc 99,999 Zn basis, PURATREM, Electronic Grade
|
Danger
|
97-5060-50G
97-5060-100G
100197-982EA
6904.25
USD
100197-982
100203-648
Dimethylzinc 99,999 Zn basis, PURATREM, Electronic Grade
Dimethylzinc
Metal Alkyls, Volatile Precursors for CVD
Formula:
C₂H₆Zn MW: 95.478 g/mol Boiling Pt: 46 °C Melting Pt: –42 °C Density: 1.386 (20°) Flash Pt: –1 °F |
MDL Number:
MFCD00014854 CAS Number: 544-97-8 UN: 3394 ADR: 4.2 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...