CleanMax® Clean Sterile Disposable Boot Covers, Lakeland Industries
Supplier: Lakeland
CTL903CSP-SMMD CTL903CSP-LGXL CTL903CSP-2X
76193-234CS 944.87 USD
76193-234 76193-236 76193-238
CleanMax® Clean Sterile Disposable Boot Covers, Lakeland Industries
Disposable Garments
Clean and Sterile disposable boot covers are individually double-bagged in pairs and folded to promote easier gowning with less excursions.
  • Clean and sterile in silicone-free environment
  • Latex-free
  • Tunneled elastic top
  • 19" High
  • Non-skid PVC sole
  • Individually double-bagged in pairs
  • Gamma radiation indicator dots on each package
CleanMax® cleanroom garments provide comfort, quality, and protection.
All these apparel provides chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunneled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
Certificaciones: Compatible for ISO Class 5 – 8 Cleanrooms and all Controlled Environments.