Magid® CutMaster® Medium Weight Spectra®/Fiberglass Blend Gloves, Magid
Supplier: Magid Glove
SP1036G-7 SP1036G-9 SP1036G-10
69100-531EA 26.52 USD
69100-531 69100-535 69100-537
Magid® CutMaster® Medium Weight Spectra®/Fiberglass Blend Gloves, Magid
Gloves
These medium weight work gloves are manufactured from a yarn blended with composite fiberglass and Spectra, one of the highest performance fibers.
  • Low lint construction minimizes product contamination
  • Dark coloration hides dirt and extends service life
  • Reversible design allows for longer wear
  • Gloves can be laundered to increase wear time
These medium weight gloves offers protection and are reversible for longer wear. Ideal glove for work in industries.
A durable and lightweight polyethylene fiber produced using a gel-spinning process, Spectra is the cutting edge in cut-resistance, and is up to 40% stronger than other aramid fibers.
Certificaciones: ANSI Level A7.