Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE)

Supplier: Hytest Ltd

AS-81006 AS-81006-100 AS-81006-1000
103010-768EA 135.92 USD InStock
103010-768 103010-770 103010-772
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE)
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE)
Although FITC reagents have been more often used to prepare fluoresceinated bioconjugates, the low stability of FITC bioconjugates makes some researchers use amine-reactive succinimidyl esters of carboxyfluorescein (commonly called FAM) in bioconjugations. FAM reagents give carboxamides that are more resistant to hydrolysis. We have shown that FAM reagents require less stringent reaction conditions and give better conjugation yields, and the resulted conjugates have superior stability. We noted that FITC labeled peptides tend to deteriorate more quickly than the corresponding FAM conjugates.

  • Spectral properties: Abs/Em = 494/519 nm
  • Solvent system: DMF or DMSO
  • Appearance: Orange Powder
  • Solubility: 1 mmol in 1ml DMF
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: Blue ice


Formula: C₂₅H₁₅NO₉
Storage Temperature: Freezer
MDL Number: MFCD04119713
CAS Number: 117548-22-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty