Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-LAMP2 Rabbit Monoclonal Antibody [clone:141] FITC (Fluorescein Isothiocyanate)

Supplier: Sino Biological
50791-R141-F-25 50791-R141-F-100
103630-162EA 70.7 USD InStock
103630-162 103632-534
Anti-LAMP2 Rabbit Monoclonal Antibody [clone:141] FITC (Fluorescein Isothiocyanate)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LAMP2/CD107b (rM LAMP2/CD107b; Catalog#50791-M08H; P17047-1; Leu26-Asn379) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: LAMP2
Clonality: Monoclonal
Clone: Clone: 141
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty